Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin A12 Antibody (1C4), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP10434A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP10434A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP10434A94 Supplier Novus Biologicals Supplier No. NBP104354AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Serpin A12 Monoclonal specifically detects Serpin A12 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Serpin A12
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1C4
Conjugate Alexa Fluor 594
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias antitrypsin), member 12, member 12, OL-64Visceral adipose-specific serpin, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serpin A12, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Vaspin, Visceral adipose tissue-derived serine protease inhibitor
Gene Symbols SERPINA12
Host Species Mouse
Immunogen Recombinant human Vaspin (21-414aa) purified from E. coli (NM_173850).
Quantity 0.1 mL
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 145264
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.