Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin A12 Antibody (VP63), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB172363 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB172363 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB172363 Supplier Novus Biologicals Supplier No. NBP280041R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Serpin A12 Monoclonal antibody specifically detects Serpin A12 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)

Specifications

Antigen Serpin A12
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone VP63
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias antitrypsin), member 12, member 12, OL-64Visceral adipose-specific serpin, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serpin A12, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Vaspin, Visceral adipose tissue-derived serine protease inhibitor
Host Species Mouse
Immunogen Recombinant human Serpin A12.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 145264
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.