Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin A3/alpha 1-Antichymotrypsin Antibody (006), FITC, Novus Biologicals™
SDP

Catalog No. NB066767 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB066767 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB066767 Supplier Novus Biologicals Supplier No. NBP289404F
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Serpin A3/alpha 1-Antichymotrypsin Monoclonal specifically detects Serpin A3/alpha 1-Antichymotrypsin in Human samples. It is validated for ELISA, Sandwich ELISA Detection.

Specifications

Antigen Serpin A3/alpha 1-Antichymotrypsin
Applications ELISA
Classification Monoclonal
Clone 6
Conjugate FITC
Dilution ELISA, Sandwich ELISA Detection
Formulation PBS
Gene Alias AACTMGC88254, ACTGIG25, alpha-1-antichymotrypsin, antitrypsin), member 3, Cell growth-inhibiting gene 24/25 protein, growth-inhibiting protein 24, growth-inhibiting protein 25, member 3, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 3, Serpin A3, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin)
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin A3/alpha 1-Antichymotrypsin (NP_001076.2; Met 1-Ala 423).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 12
Test Specificity No cross-reactivity in ELISA with: Human SerpinA7 Human SerpinA5 Human SerpinC1 Human SerpinD1 Human SerpinF2
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.