Learn More
Serpin A3/alpha 1-Antichymotrypsin Antibody (AACT/1451), Alexa Fluor™ 750, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Serpin A3/alpha 1-Antichymotrypsin |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | AACT/1451 |
| Conjugate | Alexa Fluor 750 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | AACTMGC88254, ACTGIG25, alpha-1-antichymotrypsin, antitrypsin), member 3, Cell growth-inhibiting gene 24/25 protein, growth-inhibiting protein 24, growth-inhibiting protein 25, member 3, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 3, Serpin A3, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin) |
| Gene Symbols | SERPINA3 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.