Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin A5/Protein C Inhibitor Antibody (210326), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB297210 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297210 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB297210 Supplier Novus Biologicals Supplier No. FAB1266L
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Serpin A5/Protein C Inhibitor Monoclonal antibody specifically detects Serpin A5/Protein C Inhibitor in Human samples. It is validated for Western Blot, Immunoprecipitation, Flow Cytometry, CyTOF

Specifications

Antigen Serpin A5/Protein C Inhibitor
Applications Western Blot, Immunoprecipitation, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 210326
Conjugate DyLight 550
Dilution Western Blot, Immunoprecipitation, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Acrosomal serine protease inhibitor, antitrypsin), member 5, member 5, PAI-3, PAI3PLANH3, PCIplasminogen activator inhibitor III, Plasminogen activator inhibitor 3, plasminogen activator inhibitor-3, PROCIplasma serine protease inhibitor, Protein C inhibitor, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A5, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), SERPNA5
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Serpin A5/Protein C, His20-Pro406, Accession # P05154
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5104
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.