Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin A6/Cortisol Binding Globulin Antibody (014), PE, Novus Biologicals™
SDP

Catalog No. NB332850 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB332850 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB332850 Supplier Novus Biologicals Supplier No. NBP290529PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Serpin A6/Cortisol Binding Globulin Monoclonal antibody specifically detects Serpin A6/Cortisol Binding Globulin in Mouse samples. It is validated for ELISA

Specifications

Antigen Serpin A6/Cortisol Binding Globulin
Applications ELISA
Classification Monoclonal
Clone 014
Conjugate PE
Dilution ELISA
Formulation PBS
Gene Alias antitrypsin), member 6, CBGcorticosteroid-binding globulin, corticosteroid binding globulin, member 6, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A6, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Transcortin
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse Serpin A6/Cortisol Binding Globulin (Uniprot#: Q06770, Met1-Ala397)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 866
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.