Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin A8/Angiotensinogen Antibody (001), PE, Novus Biologicals™
SDP

Catalog No. NB332799 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB332799 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB332799 Supplier Novus Biologicals Supplier No. NBP290541PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Serpin A8/Angiotensinogen Monoclonal antibody specifically detects Serpin A8/Angiotensinogen in Mouse samples. It is validated for Western Blot

Specifications

Antigen Serpin A8/Angiotensinogen
Applications Western Blot
Classification Monoclonal
Clone 001
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias alpha-1 antiproteinase, antitrypsin, angiotensin II, angiotensinogen, angiotensinogen (serpin peptidase inhibitor, clade A, member 8), ANHU, FLJ92595, FLJ97926, pre-angiotensinogen, serine (or cysteine) proteinase inhibitor, Serpin A8, SERPINA8angiotensin I
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse Serpin A8/Angiotensinogen (Uniprot#: P11859, Met1-Val477)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 183
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.