Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin A8/Angiotensinogen Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB053052 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB053052 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB053052 Supplier Novus Biologicals Supplier No. NBP298259FR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Serpin A8/Angiotensinogen Polyclonal specifically detects Serpin A8/Angiotensinogen in Mouse samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Serpin A8/Angiotensinogen
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 680
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias alpha-1 antiproteinase, antitrypsin, angiotensin II, angiotensinogen, angiotensinogen (serpin peptidase inhibitor, clade A, member 8), ANHU, FLJ92595, FLJ97926, pre-angiotensinogen, serine (or cysteine) proteinase inhibitor, Serpin A8, SERPINA8angiotensin I
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Serpin A8/Angiotensinogen (P11859; Met 1-Val 477).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 183
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.