Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin A8/Angiotensinogen Rabbit anti-Human, Clone: 1, Novus Biologicals™
SDP

Catalog No. NB020954 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB020954 50 μg
NB020953 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB020954 Supplier Novus Biologicals Supplier No. NBP28973650UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Serpin A8/Angiotensinogen Monoclonal specifically detects Serpin A8/Angiotensinogen in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen Serpin A8/Angiotensinogen
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 1
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunoprecipitation 4-6 ul/mg of lysate
Formulation 0.2 um filtered solution in PBS
Gene Alias alpha-1 antiproteinase, antitrypsin, angiotensin II, angiotensinogen, angiotensinogen (serpin peptidase inhibitor, clade A, member 8), ANHU, FLJ92595, FLJ97926, pre-angiotensinogen, serine (or cysteine) proteinase inhibitor, Serpin A8, SERPINA8angiotensin I
Host Species Rabbit
Immunogen Recombinant Human Serpin A8/Angiotensinogen protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 183
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.