Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin B3/SCCA1 Antibody (CPTC-SERPINB3-2) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB119262 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119262 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119262 Supplier Novus Biologicals Supplier No. NBP308536
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Serpin B3/SCCA1 Monoclonal specifically detects Serpin B3/SCCA1 in Human samples. It is validated for Western Blot, Protein Array.

Specifications

Antigen Serpin B3/SCCA1
Applications Western Blot, Peptide Array
Classification Monoclonal
Clone CPTC-SERPINB3-2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1-2 ug/mL, Protein Array
Formulation 10mM PBS
Gene Alias HsT1196, Protein T4-A, SCC, SCCA, SCCA1SCCA-1, SCCA-PD, serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3, serpin B3, serpin peptidase inhibitor, clade B (ovalbumin), member 3, Squamous cell carcinoma antigen 1, T4-A
Host Species Mouse
Immunogen Recombinant human full-length Serpin B3/SCCA1 protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6317
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.