Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin B3/SCCA1 Antibody (OTI1A12), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP274109FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274109FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274109FR Supplier Novus Biologicals Supplier No. NBP274109FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Serpin B3/SCCA1 Monoclonal specifically detects Serpin B3/SCCA1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Serpin B3/SCCA1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI1A12
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Alias HsT1196, Protein T4-A, SCC, SCCA, SCCA1SCCA-1, SCCA-PD, serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3, serpin B3, serpin peptidase inhibitor, clade B (ovalbumin), member 3, Squamous cell carcinoma antigen 1, T4-A
Gene Symbols SERPINB3
Host Species Mouse
Immunogen Full length recombinant protein of human Serpin B3/SCCA1 (NP_008850) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6317.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.