Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin B5/Maspin Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB11097377Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11097377Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11097377Y Supplier Novus Biologicals Supplier No. NB11097377AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Serpin B5/Maspin Polyclonal specifically detects Serpin B5/Maspin in Human samples. It is validated for Western Blot, ICC/IF (Negative), Knockdown Validated.

Specifications

Antigen Serpin B5/Maspin
Applications Western Blot, Immunocytochemistry, KnockDown
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, ICC/IF (Negative), Knockdown Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P36952
Gene Alias maspin, Peptidase inhibitor 5, PI-5, PI5protease inhibitor 5 (maspin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 5, serpin B5, serpin peptidase inhibitor, clade B (ovalbumin), member 5
Gene Symbols SERPINB5
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human MASPIN (within residues 100-200). [Swiss-Prot# P36952]
Molecular Weight of Antigen 40 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5268
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.