Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin B5/Maspin Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB338945 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338945 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338945 Supplier Novus Biologicals Supplier No. NB11097377PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Serpin B5/Maspin Polyclonal antibody specifically detects Serpin B5/Maspin in Human samples. It is validated for Western Blot, ICC/IF (Negative), KnockDown

Specifications

Antigen Serpin B5/Maspin
Applications Western Blot, KnockDown
Classification Polyclonal
Conjugate PE
Dilution Western Blot, ICC/IF (Negative), Knockdown Validated
Formulation PBS
Gene Alias maspin, Peptidase inhibitor 5, PI-5, PI5protease inhibitor 5 (maspin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 5, serpin B5, serpin peptidase inhibitor, clade B (ovalbumin), member 5
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human MASPIN (within residues 100-200). [Swiss-Prot# P36952]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5268
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.