Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin B5/Maspin Antibody (SERPINB5/4975), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB187471 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB187471 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB187471 Supplier Novus Biologicals Supplier No. NBP314136AF594
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Serpin B5/Maspin Monoclonal antibody specifically detects Serpin B5/Maspin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Serpin B5/Maspin
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SERPINB5/4975
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias maspin, Peptidase inhibitor 5, PI-5, PI5protease inhibitor 5 (maspin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 5, serpin B5, serpin peptidase inhibitor, clade B (ovalbumin), member 5
Host Species Mouse
Immunogen Recombinant fragment of human Serpin B5/Maspin protein (around aa1-200) (exact sequence is proprietary) (Uniprot: P36952)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5268
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.