Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin B6 Antibody (309), PerCP, Novus Biologicals™
SDP

Catalog No. NB316763 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB316763 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB316763 Supplier Novus Biologicals Supplier No. NBP290150PCP
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Serpin B6 Monoclonal antibody specifically detects Serpin B6 in Human samples. It is validated for ELISA

Specifications

Antigen Serpin B6
Applications ELISA
Classification Monoclonal
Clone 309
Conjugate PerCP
Dilution ELISA
Formulation PBS
Gene Alias CAPMGC111370, Cytoplasmic antiproteinase, deafness, autosomal recessive 91, DFNB91, Peptidase inhibitor 6, PI-6, PI6MSTP057, Placental thrombin inhibitor, protease inhibitor 6 (placental thrombin inhibitor), PTIDKFZp686I04222, serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 6, serpin B6, serpin peptidase inhibitor, clade B (ovalbumin), member 6, SPI3
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin B6 (Accession#: AAB30320.1; Asp 2-Pro 376).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5269
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.