Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin C1/Antithrombin-III Antibody (OTI8D5), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP274113C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274113C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274113C Supplier Novus Biologicals Supplier No. NBP274113C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Serpin C1/Antithrombin-III Monoclonal specifically detects Serpin C1/Antithrombin-III in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen Serpin C1/Antithrombin-III
Applications Flow Cytometry
Classification Monoclonal
Clone OTI8D5
Conjugate DyLight 650
Dilution Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AT3antithrombin-III, ATIIIantithrombin III, MGC22579, serine (or cysteine) proteinase inhibitor, clade C (antithrombin), member 1, serine-cysteine proteinase inhibitor clade C member 1, Serpin C1, serpin peptidase inhibitor, clade C (antithrombin), member 1
Gene Symbols SERPINC1
Host Species Mouse
Immunogen Full length recombinant protein of human Serpin C1/Antithrombin-III (NP_000479) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 462.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.