Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin C1/Antithrombin-III Antibody (OTI8D5), Novus Biologicals™
SDP

Catalog No. NBP147964 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147964 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP147964 Supplier Novus Biologicals Supplier No. NBP147964
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Serpin C1/Antithrombin-III Monoclonal specifically detects Serpin C1/Antithrombin-III in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen Serpin C1/Antithrombin-III
Applications Flow Cytometry
Classification Monoclonal
Clone OTI8D5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:100
Gene Alias AT3antithrombin-III, ATIIIantithrombin III, MGC22579, serine (or cysteine) proteinase inhibitor, clade C (antithrombin), member 1, serine-cysteine proteinase inhibitor clade C member 1, Serpin C1, serpin peptidase inhibitor, clade C (antithrombin), member 1
Gene Symbols SERPINC1
Host Species Mouse
Immunogen Full length human recombinant protein of human SERPINC1 (NP_000479) produced in HEK293T cell.
Molecular Weight of Antigen 52.6 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 462
Test Specificity This antibody is specific for Homo sapiens serpin peptidase inhibitor, clade C (antithrombin), member 1 (SERPINC1).
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.