Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin D1/Heparin Cofactor II Antibody (008), PerCP, Novus Biologicals™
SDP

Catalog No. NB307781 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB307781 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB307781 Supplier Novus Biologicals Supplier No. NBP289392PCP
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Serpin D1/Heparin Cofactor II Monoclonal antibody specifically detects Serpin D1/Heparin Cofactor II in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Serpin D1/Heparin Cofactor II
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 008
Conjugate PerCP
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias HC2LS2, HCF2clade D (heparin cofactor), member 1, HC-II, Heparin cofactor II, HLS2HCII, leuserpin 2, Protease inhibitor leuserpin-2, Serpin D1, serpin peptidase inhibitor, clade D (heparin cofactor), member 1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin D1/Heparin Cofactor II (Accession#: NP_000176.2; Met1-Ser499).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 3053
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.