Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin D1/Heparin Cofactor II Antibody (25), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB039296 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB039296 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB039296 Supplier Novus Biologicals Supplier No. NBP290429AF647
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Serpin D1/Heparin Cofactor II Monoclonal specifically detects Serpin D1/Heparin Cofactor II in Mouse samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Serpin D1/Heparin Cofactor II
Applications Western Blot, ELISA
Classification Monoclonal
Clone 25
Conjugate Alexa Fluor 647
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias HC2LS2, HCF2clade D (heparin cofactor), member 1, HC-II, Heparin cofactor II, HLS2HCII, leuserpin 2, Protease inhibitor leuserpin-2, Serpin D1, serpin peptidase inhibitor, clade D (heparin cofactor), member 1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Serpin D1/Heparin Cofactor II (NP_032249.3; Met 1-Ser 478).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 3053
Test Specificity No cross-reactivity in ELISA with: Human SerpinA1/ A1AT Human SerpinA3/ AACT Human SerpinA5/ PROCI Mouse SerpinF2/ AAP
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.