Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin D1/Heparin Cofactor II Rabbit anti-Mouse, Clone: 25, Novus Biologicals™
SDP

Catalog No. NB022312 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB022312 50 μg
NB022311 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB022312 Supplier Novus Biologicals Supplier No. NBP29042950UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Serpin D1/Heparin Cofactor II Monoclonal specifically detects Serpin D1/Heparin Cofactor II in Mouse samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen Serpin D1/Heparin Cofactor II
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 25
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunoprecipitation 0.2-1 ul/mg of lysate
Formulation 0.2 um filtered solution in PBS
Gene Alias HC2LS2, HCF2clade D (heparin cofactor), member 1, HC-II, Heparin cofactor II, HLS2HCII, leuserpin 2, Protease inhibitor leuserpin-2, Serpin D1, serpin peptidase inhibitor, clade D (heparin cofactor), member 1
Host Species Rabbit
Immunogen Recombinant Mouse Serpin D1/Heparin Cofactor II protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3053
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.