Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin E1/PAI-1 Antibody (10C3), Novus Biologicals™
SDP

Catalog No. NB298322 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298322 50 μL
NB298323 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298322 Supplier Novus Biologicals Supplier No. NBP32632750UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Serpin E1/PAI-1 Monoclonal antibody specifically detects Serpin E1/PAI-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen Serpin E1/PAI-1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 10C3
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias Endothelial plasminogen activator inhibitor, PAI1PAI-1, PAISerpin E1, PLANH1plasminogen activator inhibitor 1, serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 1, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 1
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Serpin E1/PAI-1 [UniProt P05121]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5054
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.