Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin E1/PAI-1 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP119773A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP119773A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP119773A30 Supplier Novus Biologicals Supplier No. NBP119773AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Serpin E1/PAI-1 Polyclonal specifically detects Serpin E1/PAI-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Serpin E1/PAI-1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Endothelial plasminogen activator inhibitor, PAI1PAI-1, PAISerpin E1, PLANH1plasminogen activator inhibitor 1, serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 1, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 1
Gene Symbols SERPINE1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human PAI1/Serpine 1 protein (between residues 300-400) [UniProt P05121]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5054
Test Specificity PAI-1 (N315) pAb detects endogenous levels of PAI-1 protein.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.