Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin E1/PAI-1 Mouse anti-Human, Clone: I-1, Novus Biologicals™
SDP

Catalog No. NB019308 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019308 0.025 mL
NBP22350901 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019308 Supplier Novus Biologicals Supplier No. NBP2235090.025ML

Mouse Monoclonal Antibody

Serpin E1/PAI-1 Monoclonal specifically detects Serpin E1/PAI-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoaffinity Purification.

Specifications

Antigen Serpin E1/PAI-1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone I-1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500
Formulation 10mM Phosphate (pH 7.4) and 0.14M NaCl with 15mM Sodium Azide
Gene Alias Endothelial plasminogen activator inhibitor, PAI1PAI-1, PAISerpin E1, PLANH1plasminogen activator inhibitor 1, serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 1, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 1
Gene Symbols SERPINE1
Host Species Mouse
Immunogen Partiallty purified human PAI-1 (6)
Purification Method Protein A or G purified
Quantity 0.025 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5054
Test Specificity Clone I-1 is specific for human PAI-1. No cross-reactivity is observed with PAI-2 as tested by ELISA and immunoblotting. No reactivity is observed with any other human plasma protein when tested by immunoblotting.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.