Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin E1/PAI-1 Mouse anti-Human, Clone: I-1, Novus Biologicals™

Mouse Monoclonal Antibody
$182.00 - $401.00
Specifications
Antigen | Serpin E1/PAI-1 |
---|---|
Clone | I-1 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500 |
Applications | ELISA, Immunohistochemistry, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB019308
![]() |
Novus Biologicals
NBP2235090.025ML |
0.025 mL |
Each for $182.00
|
|
|||||
NBP22350901
![]() |
Novus Biologicals
NBP2235090.1ML |
0.1 mL |
Each for $401.00
|
|
|||||
Description
Serpin E1/PAI-1 Monoclonal specifically detects Serpin E1/PAI-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoaffinity Purification.Specifications
Serpin E1/PAI-1 | |
1 mg/mL | |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Endothelial plasminogen activator inhibitor, PAI1PAI-1, PAISerpin E1, PLANH1plasminogen activator inhibitor 1, serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 1, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 1 | |
SERPINE1 | |
IgG1 | |
Protein A or G purified | |
Clone I-1 is specific for human PAI-1. No cross-reactivity is observed with PAI-2 as tested by ELISA and immunoblotting. No reactivity is observed with any other human plasma protein when tested by immunoblotting. |
I-1 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500 | |
Monoclonal | |
Purified | |
Cancer | |
10mM Phosphate (pH 7.4) and 0.14M NaCl with 15mM Sodium Azide | |
5054 | |
Partiallty purified human PAI-1 (6) | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title