Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin E1/PAI-1 Rabbit anti-Human, Clone: 4, Novus Biologicals™
SDP

Catalog No. NB020270 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB020270 50 μg
NB020269 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB020270 Supplier Novus Biologicals Supplier No. NBP28939350UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Serpin E1/PAI-1 Monoclonal specifically detects Serpin E1/PAI-1 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.

Specifications

Antigen Serpin E1/PAI-1
Applications ELISA
Classification Monoclonal
Clone 4
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias Endothelial plasminogen activator inhibitor, PAI1PAI-1, PAISerpin E1, PLANH1plasminogen activator inhibitor 1, serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 1, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 1
Host Species Rabbit
Immunogen Recombinant Human Serpin E1/PAI-1 Protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5054
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.