Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin E2/PN1 Antibody (OTI2C9), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP274117V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274117V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274117V Supplier Novus Biologicals Supplier No. NBP274117V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Serpin E2/PN1 Monoclonal specifically detects Serpin E2/PN1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen Serpin E2/PN1
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI2C9
Conjugate DyLight 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias GDNserine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 2, glia-derived nexin, glial-derived neurite promoting factor, Peptidase inhibitor 7, PI-7, PI7DKFZp686A13110, PN-1, PN1nexin, PNI, Protease nexin 1, Protease nexin I, Serpin E2, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 2
Gene Symbols SERPINE2
Host Species Mouse
Immunogen Full length recombinant protein of human Serpin E2/PN1 (NP_006207) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 5270.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.