Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin F1/PEDF Antibody (033), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB109301 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB109301 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB109301 Supplier Novus Biologicals Supplier No. NBP289799R

Rabbit Monoclonal Antibody

Serpin F1/PEDF Monoclonal specifically detects Serpin F1/PEDF in Human samples. It is validated for Western Blot.

Specifications

Antigen Serpin F1/PEDF
Applications Western Blot
Classification Monoclonal
Clone 33
Conjugate DyLight 550
Formulation 50 mM Sodium Borate
Gene Alias Cell proliferation-inducing gene 35 protein, EPC-1PIG35, PEDFpigment epithelium-derived factor, pigment epithelium derived factor), member 1, proliferation-inducing protein 35, serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, Serpin F1, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin F1/PEDF (NP_002606.3; Met1-Pro418).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Neuroscience, Vision
Primary or Secondary Primary
Gene ID (Entrez) 5176
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.