Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin F2/alpha 2-Antiplasmin Antibody (043), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB107297 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB107297 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB107297 Supplier Novus Biologicals Supplier No. NBP289397AF532
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Serpin F2/alpha 2-Antiplasmin Monoclonal specifically detects Serpin F2/alpha 2-Antiplasmin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Serpin F2/alpha 2-Antiplasmin
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 43
Conjugate Alexa Fluor 532
Formulation 50 mM Sodium Borate
Gene Alias AAPclade F (alpha-2 antiplasmin, Alpha-2-AP, Alpha-2-PI, Alpha-2-plasmin inhibitor, pigment epithelium derived factor), member 2, Serpin F2, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin F2/alpha 2-Antiplasmin (NP_000925.2; Met 1-Lys 491).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 5345
Test Specificity No cross-reactivity in ELISA with: Human SerpinA1 Human SerpinA5 Mouse SerpinD1 Mouse SerpinF1 Mouse SerpinF2
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.