Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin F2/alpha 2-Antiplasmin Antibody (RM0324-2H63), FITC, Novus Biologicals™
SDP

Catalog No. NB361598 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361598 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361598 Supplier Novus Biologicals Supplier No. NBP211890F
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Serpin F2/alpha 2-Antiplasmin Monoclonal antibody specifically detects Serpin F2/alpha 2-Antiplasmin in Mouse samples. It is validated for Western Blot

Specifications

Antigen Serpin F2/alpha 2-Antiplasmin
Applications Western Blot
Classification Monoclonal
Clone RM0324-2H63
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias AAPclade F (alpha-2 antiplasmin, Alpha-2-AP, Alpha-2-PI, Alpha-2-plasmin inhibitor, pigment epithelium derived factor), member 2, Serpin F2, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 2
Host Species Rat
Immunogen Mouse recombinant Serpin F2/alpha 2-Antiplasmin
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 5345
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.