Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin G1/C1 Inhibitor Antibody (18), Biotin, Novus Biologicals™
SDP

Catalog No. NB109031 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB109031 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB109031 Supplier Novus Biologicals Supplier No. NBP289738B
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Serpin G1/C1 Inhibitor Monoclonal specifically detects Serpin G1/C1 Inhibitor in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA Detection.

Specifications

Antigen Serpin G1/C1 Inhibitor
Applications Western Blot, ELISA
Classification Monoclonal
Clone 18
Conjugate Biotin
Formulation PBS
Gene Alias C1IN, C1INH, C1NH, HAE1, HAE2, SERPING1 serpin peptidase inhibitor, clade G (C1 inhibitor), member 1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin G1/C1 Inhibitor (NP_000053.2; Met 1-Ala 500).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 710
Test Specificity No cross-reactivity in ELISA with: Human SerpinA1/ A1AT Human SerpinA3/ SERPINA3/ AACT Human SerpinA5/ PROCI Human SerpinC1/SERPINC1/ Antithrombin-III Human SerpinD1/ SERPIND1/ HC2 Human SerpinF1/ SERPINF1/ PEDF Human SerpinF2/ SERPINF2 Human SerpinI1/ SERPINI1/ Protease inhibitor 12
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.