Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin G1/C1 Inhibitor Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NB121239 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB121239 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB121239 Supplier Novus Biologicals Supplier No. NBP311027IR
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Serpin G1/C1 Inhibitor Polyclonal antibody specifically detects Serpin G1/C1 Inhibitor in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen Serpin G1/C1 Inhibitor
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias C1IN, C1INH, C1NH, HAE1, HAE2, SERPING1 serpin peptidase inhibitor, clade G (C1 inhibitor), member 1
Host Species Sheep
Immunogen Human Serpin G1/C1 Inhibitor
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 710
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.