Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin I1/Neuroserpin Antibody (009), HRP, Novus Biologicals™
SDP

Catalog No. NB077275 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB077275 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB077275 Supplier Novus Biologicals Supplier No. NBP290746H
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Serpin I1/Neuroserpin Monoclonal specifically detects Serpin I1/Neuroserpin in Mouse samples. It is validated for ELISA.

Specifications

Antigen Serpin I1/Neuroserpin
Applications ELISA
Classification Monoclonal
Clone 9
Conjugate HRP
Formulation PBS
Gene Alias DKFZp781N13156, neuroserpin, Peptidase inhibitor 12, PI-12, PI12Serpin I1, serine (or cysteine) proteinase inhibitor, clade I (neuroserpin), member 1, serpin peptidase inhibitor, clade I (neuroserpin), member 1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Serpin I1/Neuroserpin (O35684; Met1-Leu410).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 5274
Test Specificity No cross-reactivity in ELISA with: Human SerpinA1 Human SerpinA3 Human SerpinA5 Human SerpinA12 Human SerpinB6 Human SerpinB9 Human SerpinC1 Human SerpinD1 Human SerpinF2 Human SerpinG1 Human SerpinI1 Human SerpinI2
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.