Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin I2 Antibody (018), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB067773 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB067773 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB067773 Supplier Novus Biologicals Supplier No. NBP289740AF488
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Serpin I2 Monoclonal specifically detects Serpin I2 in Human samples. It is validated for Flow Cytometry, ELISA.

Specifications

Antigen Serpin I2
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 18
Conjugate Alexa Fluor 488
Dilution Flow Cytometry, ELISA
Formulation 50mM Sodium Borate
Gene Alias MEPIpancpin, Myoepithelium-derived serine protease inhibitor, Pancpin, Pancreas-specific protein TSA2004, Peptidase inhibitor 14, PI-14, PI14clade I (neuroserpin), member 2, protease inhibitor 14, serine (or cysteine) proteinase inhibitor, clade I (pancpin), member 2, serpin I2, serpin peptidase inhibitor, clade I (pancpin), member 2, SERPIN12
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin I2 (NP_006208.1; Met 1-Leu 405).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5276
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.