Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINB10 Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NB081954 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB081954 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB081954 Supplier Novus Biologicals Supplier No. NBP298324IR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SERPINB10 Polyclonal specifically detects SERPINB10 in Mouse samples. It is validated for ELISA.

Specifications

Antigen SERPINB10
Applications ELISA
Classification Polyclonal
Conjugate DyLight 755
Formulation 50 mM Sodium Borate
Gene Alias bomapin, Peptidase inhibitor 10, PI-10, PI10protease inhibitor 10 (ovalbumin type, bomapin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 10, serpin B10, serpin peptidase inhibitor, clade B (ovalbumin), member 10
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant SERPINB10 (Q8K1K6-1; Met1-Pro397).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5273
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.