Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINB12 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10093572 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10093572 0.1 mg
NB017924 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10093572 Supplier Novus Biologicals Supplier No. NB10093572
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SERPINB12 Polyclonal specifically detects SERPINB12 in Human samples. It is validated for Western Blot.

Specifications

Antigen SERPINB12
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q96P63
Gene Alias MGC119247, MGC119248, serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 12, serpin B12, serpin peptidase inhibitor, clade B (ovalbumin), member 12, YUKOPIN
Gene Symbols SERPINB12
Host Species Rabbit
Immunogen Reacts with residues 260-275 [SHSKDNLKGLEELERK] of the human SERPINB12 protein (NM_080474)
Molecular Weight of Antigen 46.3 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89777
Test Specificity Human SERPINB12 protein
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.