Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SerpinB2 Antibody (930109), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB310727 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310727 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310727 Supplier Novus Biologicals Supplier No. FAB8550APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SerpinB2 Monoclonal specifically detects SerpinB2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen SerpinB2
Applications Flow Cytometry
Classification Monoclonal
Clone 930109
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias HsT1201, Monocyte Arg-serpin, PAI, PAI2Urokinase inhibitor, Placental plasminogen activator inhibitor, PLANH2PAI-2, plasminogen activator inhibitor 2, plasminogen activator inhibitor, type II (arginine-serpin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 2, Serpin B2, serpin peptidase inhibitor, clade B (ovalbumin), member 2
Host Species Mouse
Immunogen E. coli-derived recombinant human Serpin B2, Glu2-Ala65, Ala99-Pro415, Accession # P05120
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5055
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.