Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SerpinB2 Antibody (OTI1G3), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. N274131A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274131A647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274131A647 Supplier Novus Biologicals Supplier No. NBP274131AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SerpinB2 Monoclonal specifically detects SerpinB2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen SerpinB2
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI1G3
Conjugate Alexa Fluor 647
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias HsT1201, Monocyte Arg-serpin, PAI, PAI2Urokinase inhibitor, Placental plasminogen activator inhibitor, PLANH2PAI-2, plasminogen activator inhibitor 2, plasminogen activator inhibitor, type II (arginine-serpin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 2, Serpin B2, serpin peptidase inhibitor, clade B (ovalbumin), member 2
Gene Symbols SERPINB2
Host Species Mouse
Immunogen Full length recombinant protein of human SerpinB2 (NP_002566) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5055.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.