Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINB4 Antibody (8B8B2), PerCP, Novus Biologicals™
SDP

Catalog No. NB328154 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB328154 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB328154 Supplier Novus Biologicals Supplier No. NBP305885PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SERPINB4 Monoclonal antibody specifically detects SERPINB4 in Human samples. It is validated for ELISA

Specifications

Antigen SERPINB4
Applications ELISA
Classification Monoclonal
Clone 8B8B2
Conjugate PerCP
Dilution ELISA
Formulation PBS
Gene Alias Leupin, Peptidase inhibitor 11, PI-11, PI11clade B (ovalbumin), member 4, serpin peptidase inhibitor, clade B (ovalbumin), member 4, squamous cell carcinoma antigen 1, Squamous cell carcinoma antigen 2
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SERPINB4 (Uniprot#: P48594; Met 1-Pro 390).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 6318
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.