Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINB4 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB090850 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB090850 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB090850 Supplier Novus Biologicals Supplier No. NBP300059AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SERPINB4 Polyclonal specifically detects SERPINB4 in Human samples. It is validated for ELISA.

Specifications

Antigen SERPINB4
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50 mM Sodium Borate
Gene Alias Leupin, Peptidase inhibitor 11, PI-11, PI11clade B (ovalbumin), member 4, serpin peptidase inhibitor, clade B (ovalbumin), member 4, squamous cell carcinoma antigen 1, Squamous cell carcinoma antigen 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant SERPINB4 (P48594; Met1-Pro390).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 6318
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.