Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINB4 Antibody (OTI4C8), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP274141A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274141A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274141A30 Supplier Novus Biologicals Supplier No. NBP274141AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SERPINB4 Monoclonal specifically detects SERPINB4 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen SERPINB4
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4C8
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Leupin, Peptidase inhibitor 11, PI-11, PI11clade B (ovalbumin), member 4, serpin peptidase inhibitor, clade B (ovalbumin), member 4, squamous cell carcinoma antigen 1, Squamous cell carcinoma antigen 2
Gene Symbols SERPINB4
Host Species Mouse
Immunogen Full length recombinant protein of human SERPINB4 (NP_002965) produced in HEK293T cells.
Quantity 0.1 mL
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 6318
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.