Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SerpinB9 Antibody (7D8), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. N106439A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N106439A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N106439A94 Supplier Novus Biologicals Supplier No. NB10064359AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SerpinB9 Monoclonal specifically detects SerpinB9 in Human, Mouse (Negative), Porcine (Negative) samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen SerpinB9
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 7D8
Conjugate Alexa Fluor 594
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CAP-3, CAP3PI-9, Cytoplasmic antiproteinase 3, Peptidase inhibitor 9, PI9protease inhibitor 9 (ovalbumin type), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 9, serpin B9, serpin peptidase inhibitor, clade B (ovalbumin), member 9, serpin peptidase inhibitor, clade B, member 9
Gene Symbols SERPINB9
Host Species Mouse
Immunogen Recombinant PI-9 produced in P. pastoris.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5272
Test Specificity NB100-64359 is specific for human PI-9 (proteinase inhibitor 9), also known as SerpinB9, a 42kDa intracellular nucleocytoplasmic serpin expressed in cytotoxic lymphocytes (CTLs), natural killer (NK) cells, monocyte-derived dendritic cells (DCs), and to a lesser extent in B cells and myeloid cells. Granzyme B (grB) is a serine protease highly expressed by CTLs and NK cells, which is endocytosed by virus-infected and malignant target cells. The subsequent release of grB from the endocytic vesicles into the cytoplasm of the target cells, triggers grB-mediated apoptosis, through cleavage of various cytoplasmic or nuclear proteins. PI-9, up-regulated in response to grB production and degranulation, has been identified as a potent inhibitor of Granzyme B-mediated apoptosis, providing a vital self-protection mechanism against the premature apoptosis of CTLs and NK cells by grB, which may escape into the cytoplasm of the effector cells themselves. Clone 7D8 detects a band of approximately 42kDa under reducing conditions. Clone 7D8 is suitable for use in indirect Immunofluorescence (2).
Target Species Human, Mouse (Negative), Pig (Negative)
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.