Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sertad1 Antibody
SDP

Catalog No. NB10075447 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10075447 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10075447 Supplier Novus Biologicals Supplier No. NB10075447

Chicken Polyclonal Antibody

Sertad1 Polyclonal specifically detects Sertad1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Sertad1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS with No Preservative
Gene Accession No. Q53GC0
Gene Alias SEI-1, SEI1CDK4-binding protein p34SEI1, SERTA domain containing 1, SERTA domain-containing protein 1, Transcriptional regulator interacting with the PHD-bromodomain 1, TRIP-Br1CDK4-binding protein p34SEI
Gene Symbols SERTAD1
Host Species Chicken
Immunogen The immunogen for this antibody was a sequence from 1a.a. to 236a.a. from Sertad1(a 236a.a. long protein)
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 29950
Test Specificity Sertad1
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.