Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serum Amyloid A1 Antibody (SAA1cc), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB360103 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360103 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360103 Supplier Novus Biologicals Supplier No. NB10073071AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Serum Amyloid A1 Monoclonal specifically detects Serum Amyloid A1 in Canine, Human samples. It is validated for ELISA, Western Blot.

Specifications

Antigen Serum Amyloid A1
Applications ELISA, Western Blot
Classification Monoclonal
Clone SAA1cc
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias MGC111216, PIG4, SAA, SAA2, serum amyloid A protein, serum amyloid A1, TP53I4, tumor protein p53 inducible protein 4
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human SAA.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Biologically Active Proteins, Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6288
Target Species Canine, Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.