Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serum Amyloid A1 Antibody (SAA1cc), FITC, Novus Biologicals™
SDP

Catalog No. NB001749 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001749 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001749 Supplier Novus Biologicals Supplier No. NB10073071F200UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Serum Amyloid A1 Monoclonal specifically detects Serum Amyloid A1 in Canine, Human samples. It is validated for ELISA, Western Blot.

Specifications

Antigen Serum Amyloid A1
Applications ELISA, Western Blot
Classification Monoclonal
Clone SAA1
Conjugate FITC
Dilution ELISA
Formulation PBS
Gene Alias MGC111216, PIG4, SAA, SAA2, serum amyloid A protein, serum amyloid A1, TP53I4, tumor protein p53 inducible protein 4
Gene Symbols SAA1
Host Species Mouse
Immunogen human recombinant SAA, conjugated with carrier protein.
Quantity 200 μL
Research Discipline Alzheimers Research, Cancer, Neuroscience
Gene ID (Entrez) 6288
Test Specificity Human recombinant SAA
Target Species Canine, Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.