Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serum Response Factor SRF Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB290979 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290979 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290979 Supplier Novus Biologicals Supplier No. NBP171927APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Serum Response Factor SRF Polyclonal antibody specifically detects Serum Response Factor SRF in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen Serum Response Factor SRF
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias MCM1, serum response factor, serum response factor (c-fos serum response element-binding transcriptionfactor)
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 25 and 75 of human Serum Response Factor using the numbering given in entry NP_003122.1 (GeneID 6722).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 6722
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.