Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serum Response Factor SRF Antibody (PCRP-SRF-1F1), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB156221 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB156221 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB156221 Supplier Novus Biologicals Supplier No. NBP314187AF750
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Serum Response Factor SRF Monoclonal antibody specifically detects Serum Response Factor SRF in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen Serum Response Factor SRF
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-SRF-1F1
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias MCM1, serum response factor, serum response factor (c-fos serum response element-binding transcriptionfactor)
Host Species Mouse
Immunogen Recombinant full-length human Serum Response Factor SRF protein (Uniprot: P11831)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6722
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.