Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SESN1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP14499350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP14499350 50 μg
NBP14499310 100 μg
NB018556 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP14499350 Supplier Novus Biologicals Supplier No. NBP14499350UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SESN1 Polyclonal specifically detects SESN1 in Human, Mouse, Rat samples. It is validated for ELISA, Western Blot.

Specifications

Antigen SESN1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot >1:500, ELISA < 1:10000
Formulation Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% sodium azide
Gene Alias MGC138241, MGC142129, p53-regulated protein PA26, PA26p53 activated gene 26, SEST1p53 regulated PA26 nuclear protein, sestrin 1, sestrin-1
Gene Symbols SESN1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to unique epitope on SESN1. The SESN1 peptide was selected from N-terminal, amino acids 1-50. This peptide was covalently modified post-synthetically to achieve desired antigenicity.
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) 27244
Test Specificity This antibody is specific to SESN1
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.