Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SESN2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP14498910 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP14498910 100 μg
NBP14498950 50 μg
NB018555 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP14498910 Supplier Novus Biologicals Supplier No. NBP144989100UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SESN2 Polyclonal specifically detects SESN2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Dot Blot.

Specifications

Antigen SESN2
Applications Western Blot, ELISA, Dot Blot
Classification Polyclonal
Concentration 0.68 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:10000, Dot Blot 1:10000
Formulation Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% Sodium Azide
Gene Accession No. P58004
Gene Alias DKFZp761M0212, DKFZp761M02121, Hi95, HI95sestrin-2, SES2, SEST2hypoxia induced gene 95, sestrin 2
Gene Symbols SESN2
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 1-80 on SESN2 protein.
Molecular Weight of Antigen 54.6 kDa
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 83667
Test Specificity This antibody is specific to SESN2
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.