Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SESN3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP14498550 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP14498550 50 μg
NBP14498510 100 μg
NB018554 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP14498550 Supplier Novus Biologicals Supplier No. NBP14498550UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SESN3 Polyclonal specifically detects SESN3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen SESN3
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.50 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:10000
Formulation Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% sodium azide
Gene Accession No. P58005
Gene Alias SEST3MGC29667, sestrin 3, sestrin-3
Gene Symbols SESN3
Host Species Rabbit
Immunogen Synthetic peptides corresponding to unique epitope on SESN3. The SESN3 peptide was selected from N-terminal, amino acids 1-50. This peptide was covalently modified post-synthetically to achieve desired antigenicity.
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) 143686
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.