Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SESN3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$203.51 - $653.50
Specifications
| Antigen | SESN3 |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Target Species | Human, Mouse, Rat |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP14498550
![]() |
Novus Biologicals
NBP14498550UG |
50 μg |
Each for $394.26
|
|
|||||
NBP14498510
![]() |
Novus Biologicals
NBP144985100UG |
100 μg |
Each for $653.50
|
|
|||||
NB018554
![]() |
Novus Biologicals
NBP1449850.025MG |
0.025 mg |
Each for $203.51
|
|
|||||
Description
SESN3 Polyclonal specifically detects SESN3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
| SESN3 | |
| Unconjugated | |
| Human, Mouse, Rat | |
| SEST3MGC29667, sestrin 3, sestrin-3 | |
| SESN3 | |
| IgG |
| Polyclonal | |
| Rabbit | |
| P58005 | |
| 143686 | |
| Synthetic peptides corresponding to unique epitope on SESN3. The SESN3 peptide was selected from N-terminal, amino acids 1-50. This peptide was covalently modified post-synthetically to achieve desired antigenicity. | |
| Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title